Retort rods, with M10 thread
12,5 mm, 750 mm, Stål, M10
Katalog # NICK8632
Leverandør: NICKEL ELECTRO
Product Details & Documents
Aluminium, mild steel or stainless steel.
Bestillingsoplysninger: Retort bases are not included, they can be ordered separately under "Complementary products".
Specifikationer
- Ø:12,5 mm
- Gevind:M10
- Længde:750 mm
- Materiale:Stål
Complementary Products
Produktinformation
- Katalog #BeskrivelseL×BTilgængelighedPrice
- NICK6890Retortfod, med gevind M10 × 1,5 mm, presset stål med nupret mat sort akrylmalet overflade160×100 mmLog ind for tilgængelighed1 * 1 Items - 165,00 kr
- NICK6893Retortfod, med gevind M10 × 1,5 mm, presset stål med nupret mat sort akrylmalet overflade, med 2 gevindpositioner315×200 mmLog ind for tilgængelighed1 * 1 Items - 413,00 kr
- NICK6891Retortfod, med gevind M10 × 1,5 mm, presset stål med nupret mat sort akrylmalet overflade200×125 mmLog ind for tilgængelighed1 * 1 Items - 196,00 kr
- NICK6892Retortfod, med gevind M10 × 1,5 mm, presset stål med nupret mat sort akrylmalet overflade250×160 mmLog ind for tilgængelighed1 * 1 Items - 291,00 kr
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



