Filter candles
1, 13×25 mm
Catalog # LENZ07206125
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
These cylindrical filter plugs without tube are made of borosilicate glass 3,3.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Diameter×H:13×25 mm
- Porøsitet:1
- Seq:07