Filter candles
0, 9×20 mm
Katalog # LENZ07205020
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
These cylindrical filter plugs without tube are made of borosilicate glass 3,3.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Diameter×H:9×20 mm
- Porøsitet:0
- Seq:01