Orders LinkContactUsLinkComponent
Filtertragte
Filtertragte
500 ml, 95 mm, 1
Katalog # LENZ07195158
Leverandør:   Lenz Laborglas
Filtertragte
Katalog # LENZ07195158
Leverandør:  Lenz Laborglas
Filtertragte
Filtertragte
500 ml, 95 mm, 1
Price
745,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filters disc.
Documents
Specifikationer
  • Plade Ø:
    95 mm
  • Porøsitet:
    1
  • Kapacitet:
    500 ml
  • Seq:
    13