Orders LinkContactUsLinkComponent
Filterskiver
Filterskiver
2, 40 mm
Catalog # LENZ07180240
Supplier:   Lenz Laborglas
Filterskiver
Catalog # LENZ07180240
Supplier:  Lenz Laborglas
Filterskiver
Filterskiver
2, 40 mm
Price
523,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Glass filter discs centred with unfused rim.
Documents
Specifications
  • Ø:
    40 mm
  • Porøsitet:
    2
  • Seq:
    16