Orders LinkContactUsLinkComponent
Retort bases, with M10 thread
Retort bases, with M10 thread
Retortfod, med gevind M10 × 1,5 mm, presset stål med nupret mat sort akrylmalet overflade, 250×160 mm
Catalog # NICK6892
Supplier:   NICKEL ELECTRO
Retort bases, with M10 thread
Catalog # NICK6892
Supplier:  NICKEL ELECTRO
Retort bases, with M10 thread
Retort bases, with M10 thread
Retortfod, med gevind M10 × 1,5 mm, presset stål med nupret mat sort akrylmalet overflade, 250×160 mm
Price
291,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Pressed steel finished in matt black textured acrylic paint, threaded M10 × 1,5 mm.
Ordering information: Retort rods are not included, they can be ordered separately under "Complementary products".
Specifications
  • Beskrivelse:
    Retortfod, med gevind M10 × 1,5 mm, presset stål med nupret mat sort akrylmalet overflade
  • L×B:
    250×160 mm
Complementary Products
Product Information
  • Catalog #
    Materiale
    Ø
    Længde
    Gevind
    Availability
    Price
  • NICK8631
    Stål
    12,5 mm
    600 mm
    M10
    Log in for availability
    1 * 1 Items - 237,00 kr
  • NICK8632
    Stål
    12,5 mm
    750 mm
    M10
    Log in for availability
    1 * 1 Items - 118,00 kr
  • NICK8614
    aluminium
    12 mm
    500 mm
    M10
    Log in for availability
    1 * 1 Items - 136,00 kr
  • NICK8633
    Stål
    12,5 mm
    1000 mm
    M10
    Log in for availability
    1 * 1 Items - 151,00 kr
  • NICK8634
    Stål
    10 mm
    500 mm
    M10
    Log in for availability
    1 * 1 Items - 81,90 kr
  • NICK8624
    Rustfrit stål
    10 mm
    500 mm
    M10
    Log in for availability
    1 * 1 Items - 439,00 kr