Retort bases, with M10 thread
Retortfod, med gevind M10 × 1,5 mm, presset stål med nupret mat sort akrylmalet overflade, 160×100 mm
Katalog # NICK6890
Leverandør: NICKEL ELECTRO
Product Details & Documents
Pressed steel finished in matt black textured acrylic paint, threaded M10 × 1,5 mm.
Bestillingsoplysninger: Retort rods are not included, they can be ordered separately under "Complementary products".
Specifikationer
- Beskrivelse:Retortfod, med gevind M10 × 1,5 mm, presset stål med nupret mat sort akrylmalet overflade
- L×B:160×100 mm
Complementary Products
Produktinformation
- Katalog #MaterialeØLængdeGevindTilgængelighedPrice