Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Urglas

Urglas

Watch glass, PTFE, 76,32 mm
Katalog # KART644
Leverandør:   KARTELL
Urglas
Katalog # KART644
Leverandør:  KARTELL
Urglas
Urglas

Watch glass, PTFE, 76,32 mm

Price
141,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Made of PTFE, white.

  • Very good chemical, heat and mechanical resistance
  • Heat resistant up to +120 °C
  • For use with foodstuff

For blends or for covering vessels.

Specifikationer
  • Ø:
    76,32 mm
  • Beskrivelse:
    Watch glass, PTFE
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.