Stirring shafts
WB 16, 260 mm, 16 mm
Katalog # LENZ06300516
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
Stirring shafts made of precision calibrated glass tube.
- Ground and polished bearing surface
- Interchangeable
- Made of DURAN® tubing
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Ø:16 mm
- Model:WB 16
- Længde:260 mm
- Seq:02