Orders LinkContactUsLinkComponent
Flat flange lids with 4 necks
NS 45/40 (Centre neck), 2×29/32 angled and 1×14/23 straight NS
  LENZ06104464
Flat flange lids with 4 necks
  LENZ06104464
Flat flange lids with 4 necks
Flat flange lids with 4 necks
NS 45/40 (Centre neck), 2×29/32 angled and 1×14/23 straight NS
3.740,00 kr
1 * 1 Items
DURAN®, borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With flat laboratory flange (DN). Straight middle neck, two angled and one straight side necks with standard ground joints.
  • Hals:
    NS 45/40 (Centre neck)
  • Sidehalse:
    2×29/32 angled and 1×14/23 straight NS