Orders LinkContactUsLinkComponent
Flat flange lids, centre neck
Flat flange lids, centre neck
laboratorieflange DN 200, 1 hals, NS 45/40 (center), 165 mm
Katalog # LENZ06100506
Leverandør:   Lenz Laborglas
Flat flange lids, centre neck
Katalog # LENZ06100506
Leverandør:  Lenz Laborglas
Flat flange lids, centre neck
Flat flange lids, centre neck
laboratorieflange DN 200, 1 hals, NS 45/40 (center), 165 mm
Price
4.290,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
Documents
Specifikationer
  • Model:
    1 hals
  • med:
    laboratorieflange DN 200
  • Hals:
    NS 45/40 (center)
  • Samlet højde:
    165 mm