Flat flange lids, centre neck
laboratorieflange DN 200, 1 hals, NS 29/32 (center), 150 mm
Catalog # LENZ06100504
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Model:1 hals
- med:laboratorieflange DN 200
- Hals:NS 29/32 (center)
- Samlet højde:150 mm