Flat flange lids, centre neck
laboratorieflange DN 150, 1 hals, NS 45/40 (center), 140 mm
LENZ06100406
DURAN®, Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
- Model:1 hals
- med:laboratorieflange DN 150
- Hals:NS 45/40 (center)
- Samlet højde:140 mm