Flat flange lids, centre neck
laboratorieflange DN 150, 1 hals, NS 29/32 (center), 125 mm
Katalog # LENZ06100404
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Model:1 hals
- med:laboratorieflange DN 150
- Hals:NS 29/32 (center)
- Samlet højde:125 mm