Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Test tubes with NS stopper

Test tubes with NS stopper

150 mm, 28 mm, 24/29 NS
Katalog # LENZ05700238
Leverandør:   Lenz Laborglas
Test tubes with NS stopper
Katalog # LENZ05700238
Leverandør:  Lenz Laborglas
Test tubes with NS stopper
Test tubes with NS stopper

150 mm, 28 mm, 24/29 NS

Price
144,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

DURAN®, borosilicate glass, clear. With standard ground joint and hexagonal glass stopper.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Documents

Specifikationer
  • Udv.Ø:
    28 mm
  • Kappestr.:
    24/29 NS
  • Længde:
    150 mm
  • Seq:
    05
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.