Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Test tubes with NS stopper

Test tubes with NS stopper

180 mm, 18 mm, 14/23 NS
Katalog # LENZ05700230
Leverandør:   Lenz Laborglas
Test tubes with NS stopper
Katalog # LENZ05700230
Leverandør:  Lenz Laborglas
Test tubes with NS stopper
Test tubes with NS stopper

180 mm, 18 mm, 14/23 NS

Price
94,90 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

DURAN®, borosilicate glass, clear. With standard ground joint and hexagonal glass stopper.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

Documents

Specifikationer
  • Udv.Ø:
    18 mm
  • Kappestr.:
    14/23 NS
  • Længde:
    180 mm
  • Seq:
    07