Test tubes with NS stopper
150 mm, 18 mm, 14/23 NS
LENZ05700226
DURAN®, borosilicate glass, clear. With standard ground joint and hexagonal glass stopper.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
- Udv.Ø:18 mm
- Kappestr.:14/23 NS
- Længde:150 mm
- Seq:03



