Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Centrifugerør, ASTM

Centrifugerør, ASTM

Pæreformet, cylindrisk bund, ASTM D96, 100 ml, 160 mm, uden cap
Catalog # LENZ05675005
Supplier:   Lenz Laborglas
Centrifugerør, ASTM
Catalog # LENZ05675005
Supplier:  Lenz Laborglas
Centrifugerør, ASTM
Centrifugerør, ASTM

Pæreformet, cylindrisk bund, ASTM D96, 100 ml, 160 mm, uden cap

Price
752,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

DURAN®, borosilicate glass.

  • Withstands forces up to 700 ×g
  • Good temperature resistance
  • Good chemical resistance

With conical bottom or pear shaped.

Documents

Specifications
  • Lukkestørrelse:
    uden cap
  • Kapacitet:
    100 ml
  • Beskrivelse:
    Pæreformet, cylindrisk bund, ASTM D96
  • Længde:
    160 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.