Bottle top tilt measures
1 ml, klar
613-6251
DURAN®, borosilicate glass 3.3, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
This dispenser head is completed by a 1 litre bottle for repetitive dispensing of the same volume.
NS 29/32
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
- Kapacitet:1 ml
- Farve:klar