Orders LinkContactUsLinkComponent
Extractor heads for specific light solvents
Extractor heads for specific light solvents
500 ml
Katalog # LENZ05432158
Leverandør:   Lenz Laborglas
Extractor heads for specific light solvents
Katalog # LENZ05432158
Leverandør:  Lenz Laborglas
Extractor heads for specific light solvents
Extractor heads for specific light solvents
500 ml
Price
2.950,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific light solvents, with insert filter (porosity: 0).
Documents
Specifikationer
  • Ekstraktionskap.:
    500 ml
  • Seq:
    03