Orders LinkContactUsLinkComponent
Glass beads
Glass beads
Glass beads, solid, 8 mm
Katalog # 201-0524
Leverandør:   Lenz Laborglas
Glass beads
Katalog # 201-0524
Leverandør:  Lenz Laborglas
Glass beads
Glass beads
Glass beads, solid, 8 mm
Price
497,00 kr
Log ind for tilgængelighed
1 * 1 kg
Product Details & Documents
Solid soda-lime glass.
Specifikationer
  • Ø:
    8 mm
  • Beskrivelse:
    Glass beads, solid
  • Seq:
    06