Orders LinkContactUsLinkComponent
Distillation columns, Hempel
Distillation columns, Hempel
500 mm, 29/32 NS, 29/32 NS
Katalog # LENZ05117504
Leverandør:   Lenz Laborglas
Distillation columns, Hempel
Katalog # LENZ05117504
Leverandør:  Lenz Laborglas
Distillation columns, Hempel
Distillation columns, Hempel
500 mm, 29/32 NS, 29/32 NS
Price
1.030,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours, with cooling coil and cooling mantle. Standard ground cone and jacket. Water supplied through plastic connectors with 8 mm external Ø and GL threaded joint.
DIN 12593
Certifications: DIN 12593
Documents
Specifikationer
  • Konusstr.:
    29/32 NS
  • Kappestr.:
    29/32 NS
  • Brugbar længde:
    500 mm