Orders LinkContactUsLinkComponent
Dropping funnels, conical shape, without pressure balance
Dropping funnels, conical shape, without pressure balance
1000 ml, 29/32 NS, 29/32 NS
Katalog # 532-0050
Leverandør:   Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Katalog # 532-0050
Leverandør:  Lenz Laborglas
Dropping funnels, conical shape, without pressure balance
Dropping funnels, conical shape, without pressure balance
1000 ml, 29/32 NS, 29/32 NS
Price
1.180,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Borosilicate glass 3.3.
  • Screw thread retaining device
  • With inscription label, without graduation
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.
Documents
Specifikationer
  • Konusstr.:
    29/32 NS
  • Kappestr.:
    29/32 NS
  • Kapacitet:
    1000 ml