Orders LinkContactUsLinkComponent
Dropping funnels, conical shape, without pressure balance
50 ml, 19/26 NS, 19/26 NS
  201-0909
Dropping funnels, conical shape, without pressure balance
  201-0909
Dropping funnels, conical shape, without pressure balance
Dropping funnels, conical shape, without pressure balance
50 ml, 19/26 NS, 19/26 NS
631,00 kr
1 * 1 Items
Borosilicate glass 3.3.
  • Screw thread retaining device
  • With inscription label, without graduation
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.
  • Konusstr.:
    19/26 NS
  • Kappestr.:
    19/26 NS
  • Kapacitet:
    50 ml