Dropping funnels, conical shape, without pressure balance
50 ml, 19/26 NS, 19/26 NS
201-0909
Borosilicate glass 3.3.
- Screw thread retaining device
- With inscription label, without graduation
- Minimal thermal expansion giving relatively high resistance to temperature changes
With NS socket, NS cone and stopcock with glass or PTFE plug.
- Konusstr.:19/26 NS
- Kappestr.:19/26 NS
- Kapacitet:50 ml