Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Propper, silikonegummi

Propper, silikonegummi

0, 60,0 mm, 43,0 mm, 60,0 mm
Katalog # KART3816
Leverandør:   KARTELL
Propper, silikonegummi
Katalog # KART3816
Leverandør:  KARTELL
Propper, silikonegummi
Propper, silikonegummi

0, 60,0 mm, 43,0 mm, 60,0 mm

Price
482,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

These translucent conical stoppers are suitable for foodstuffs.

Specifikationer
  • Ø bund:
    43,0 mm
  • Ø top:
    60,0 mm
  • Ant. huller:
    0
  • Højde:
    60,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.