Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Propper, silikonegummi

0, 24,0 mm, 14,0 mm, 20,0 mm
  KART3806
 :   KARTELL
Propper, silikonegummi
  KART3806
 :  KARTELL
Propper, silikonegummi
Propper, silikonegummi

0, 24,0 mm, 14,0 mm, 20,0 mm

 
24,90 kr
   
1 * 1 Items
 

These translucent conical stoppers are suitable for foodstuffs.

 
  • Ø bund:
    14,0 mm
  • Ø top:
    20,0 mm
  • Ant. huller:
    0
  • Højde:
    24,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.