L-shaped spreaders, PS
L-shaped bacteriology loop, with special angle inclination, individually wrapped
Leverandør: KARTELL
Gå til:
Product Details & Documents
Disposable L-shaped sterile PS spreaders for bacteriology with special 100° angling for easier use. Especially handy for dispersing liquids across agar culture plates.
Emballering: Available single-wrapped or in packs of 5.
Documents
Specifikationer
- Beskrivelse:L-shaped bacteriology loop, with special angle inclination, individually wrapped
- Seq:02