Orders LinkContactUsLinkComponent
L-shaped spreaders, PS
L-shaped spreaders, PS
L-shaped bacteriology loop, with special angle inclination, individually wrapped
Katalog # KART2647
Leverandør:   KARTELL
L-shaped spreaders, PS
Katalog # KART2647
Leverandør:  KARTELL
L-shaped spreaders, PS
L-shaped spreaders, PS
L-shaped bacteriology loop, with special angle inclination, individually wrapped
Price
6.760,00 kr
Log ind for tilgængelighed
1 * 2.000 Items
Product Details & Documents
Disposable L-shaped sterile PS spreaders for bacteriology with special 100° angling for easier use. Especially handy for dispersing liquids across agar culture plates.
Emballering: Available single-wrapped or in packs of 5.
Specifikationer
  • Beskrivelse:
    L-shaped bacteriology loop, with special angle inclination, individually wrapped
  • Seq:
    02