Adaptere, NS-muffe til kugleslib
29/32 NS, S 41
LENZ01532941
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapters from spherical joint ball to ground joint socket.
- Kugle:S 41
- Top:29/32 NS