Adaptere, NS-muffe til kugleslib
29/32 NS, S 35
Catalog # LENZ01532935
Supplier: Lenz Laborglas
Jump to:
Product Details & Documents
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapters from spherical joint ball to ground joint socket.
Documents
BSE TSE Glass Columns
BSE/TSE Certificates
Specifications
- Kugle:S 35
- Top:29/32 NS