Orders LinkContactUsLinkComponent
Adaptere, NS-muffe til kugleslib
Adaptere, NS-muffe til kugleslib
29/32 NS, S 29
Katalog # LENZ01532929
Leverandør:   Lenz Laborglas
Adaptere, NS-muffe til kugleslib
Katalog # LENZ01532929
Leverandør:  Lenz Laborglas
Adaptere, NS-muffe til kugleslib
Adaptere, NS-muffe til kugleslib
29/32 NS, S 29
Price
337,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapters from spherical joint ball to ground joint socket.
Documents
Specifikationer
  • Kugle:
    S 29
  • Top:
    29/32 NS