Adaptere, NS-muffe til kugleslib
14/23 NS, S 19
Katalog # LENZ01531419
Leverandør: Lenz Laborglas
Gå til:
Product Details & Documents
DURAN®, borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapters from spherical joint ball to ground joint socket.
Documents
BSE TSE Glass Columns
BSE/TSE-certifikater
Specifikationer
- Kugle:S 19
- Top:14/23 NS