Sleeve samplers, U-D Master
Prøvekammer (type 2), 1,3 – 2,5 ml
Catalog # SAMP1500A-04
Supplier: Sampling Systems
Jump to:
Product Details & Documents
The U-D sleeve sampler was developed for accurate unit dose samples (small volume).
- Quick and easy accurate unit dose sampling
- Different volumes can be taken using the same sampler
- Simply change the tip for a different volume
Designed for use with free flowing powders and granules.
Specifications
- Volumen:1,3 – 2,5 ml
- Beskrivelse:Prøvekammer (type 2)
- Seq:05