Sleeve samplers, U-D Master
Prøvekammer, type 1, til spidser 0,1 til 1,2 ml, 0,1 – 1,2 ml
Katalog # SAMP1500A-03
Leverandør: Sampling Systems
Gå til:
Product Details & Documents
The U-D sleeve sampler was developed for accurate unit dose samples (small volume).
- Quick and easy accurate unit dose sampling
- Different volumes can be taken using the same sampler
- Simply change the tip for a different volume
Designed for use with free flowing powders and granules.
Specifikationer
- Volumen:0,1 – 1,2 ml
- Beskrivelse:Prøvekammer, type 1, til spidser 0,1 til 1,2 ml
- Seq:04