Orders LinkContactUsLinkComponent
Spore strips, Attest™
Spore strips, Attest™
Bacillus subtilis-sporestrip, ethylenoxidsterilisation
Catalog # MMMEM1264
Supplier:   3M MEDICAL
Spore strips, Attest™
Catalog # MMMEM1264
Supplier:  3M MEDICAL
Spore strips, Attest™
Spore strips, Attest™
Bacillus subtilis-sporestrip, ethylenoxidsterilisation
Price
2.360,00 kr
Log in for availability
1 * 100 Items
Product Details & Documents
3M™ Attest™ Biological Monitoring Systems are true biological indicators, in self-contained indicator capsules. They enable checks on the biological reliability and performance of each steriliser without laboratory facilities and laboratory trained personnel.
  • Self-contained, reducing the risk of false positives due to contamination
  • Colour change gives clear unambiguous, visual indication of PASS or FAIL
  • Time saving and results in 24 to 48 hours which require no interpretation
Specifications
  • Beskrivelse:
    Bacillus subtilis-sporestrip, ethylenoxidsterilisation