Engangsørepropper, Bilsom 303
Earplugs, 10 pairs/PocketPak, gul, L
Catalog # 111-2157
Supplier: HONEYWELL SAFETY
Jump to:
Product Details & Documents
Formstøbt skumpropper uden PVC, fås i to størrelser for at sikre korrekt og komfortabel pasform.
- Polyurethane foam, without cord
- Conical shape for easy insertion
- Smooth outer surface for better hygiene
- Transparent dispenser for convenience
SNR: 33 dB
EN 352-2
Certifications: EN 352-2.
Packaging: Available bulk packed, as individual pairs or as 400 pairs within the LS-400 dispenser.
Specifications
- Farve:gul
- Beskrivelse:Earplugs, 10 pairs/PocketPak
- Seq:07
- Størrelse:L
Accessories
Product Information
- Catalog #BeskrivelseFarveAvailabilityPrice
- DALL1006203-AMEar plug dispenser for use with earplugssortLog in for availability1 * 1 Items - 921,00 kr