Orders LinkContactUsLinkComponent
Engangsørepropper, Bilsom 303
Engangsørepropper, Bilsom 303
Earplugs, 10 pairs/PocketPak, gul, L
Catalog # 111-2157
Supplier:   HONEYWELL SAFETY
Engangsørepropper, Bilsom 303
Catalog # 111-2157
Supplier:  HONEYWELL SAFETY
Engangsørepropper, Bilsom 303
Engangsørepropper, Bilsom 303
Earplugs, 10 pairs/PocketPak, gul, L
Price
kr446.00
Log in for availability
1 * 200 Par
Product Details & Documents
Formstøbt skumpropper uden PVC, fås i to størrelser for at sikre korrekt og komfortabel pasform.
  • Polyurethane foam, without cord
  • Conical shape for easy insertion
  • Smooth outer surface for better hygiene
  • Transparent dispenser for convenience
SNR: 33 dB
EN 352-2
Certifications: EN 352-2.
Packaging: Available bulk packed, as individual pairs or as 400 pairs within the LS-400 dispenser.
Specifications
  • Farve:
    gul
  • Beskrivelse:
    Earplugs, 10 pairs/PocketPak
  • Seq:
    07
  • Størrelse:
    L
Accessories
Product Information
  • Catalog #
    Beskrivelse
    Farve
    Availability
    Price
  • DALL1006203-AM
    Ear plug dispenser for use with earplugs
    sort
    Log in for availability
    1 * 1 Items - 921,00 kr