Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Omrøreraksler og -blade

Omrøreraksler og -blade

Shafts for blades, 300 mm, 6 mm
Katalog # COWI005.530.1
Leverandør:   COWIE
Omrøreraksler og -blade
Katalog # COWI005.530.1
Leverandør:  COWIE
Omrøreraksler og -blade
Omrøreraksler og -blade

Shafts for blades, 300 mm, 6 mm

Price
395,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

PTFE. Interchangeable stirring shafts and blades, please order shafts and blades separately.

Specifications
  • Ø:
    6 mm
  • Model:
    Shafts for blades
  • Længde:
    300 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.