Orders LinkContactUsLinkComponent
Magnetic stirring bars, plain, PTFE
Magnetic stirring bars, plain, PTFE
60 mm, 10 mm
Katalog # COWI001.260
Leverandør:   COWIE
Magnetic stirring bars, plain, PTFE
Katalog # COWI001.260
Leverandør:  COWIE
Magnetic stirring bars, plain, PTFE
Magnetic stirring bars, plain, PTFE
60 mm, 10 mm
Price
49,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Plain PTFE magnetic stirring bars have similar performance to cylindrical bars, but provide more turbulence at low speeds.
Specifikationer
  • Ø:
    10 mm
  • Længde:
    60 mm
  • Seq:
    10