Orders LinkContactUsLinkComponent
General purpose funnels, PP/PE
8 mm, 60 mm
  BURK9602-0060
 :   BURKLE
General purpose funnels, PP/PE
  BURK9602-0060
 :  BURKLE
General purpose funnels, PP/PE
General purpose funnels, PP/PE
8 mm, 60 mm
14,10 kr
1 * 1 Items
All-purpose funnels with suspension eyelet for safe filling of containers.
  • Tragt udv. Ø:
    60 mm
  • Seq:
    05
  • Stilk udv. Ø:
    8 mm
Accessories
  •  
    Beskrivelse
  • BURK9602-0251
    Sieve for barrel funnels Ø 50 mm
    1 * 1 Items - 42,50 kr