Saponin
Glass bottle for solids, 25 g
A18820.14
CAS No.: 8047-15-2 Physical Form: Powder Appearance: White to pale yellow Melting Point: 158° dec. Merck Reference: 14,8365 MDL No.: MFCD00081981
A glycoside used to permeabilize cellular membranes
- :8047-15-2
- :MFCD00081981
- :14,8365
- :158 °C (dec.)
- :1,015…1.02 g/cm³ (20 °C)
- :Stue temperatur
- :232-462-6
- Pakning:Glass bottle for solids
- Pakn:25 g