Orders LinkContactUsLinkComponent
Testkits, chlortest, til Spectroquant®, Supelco®
Testkits, chlortest, til Spectroquant®, Supelco®
Klorreagens Cl₂-2*4, -, Prove, Nova 60, Multy, Move, Picco, 400 Tests
Katalog # 1.00087.0001
Leverandør:   Merck
Testkits, chlortest, til Spectroquant®, Supelco®
Katalog # 1.00087.0001
Leverandør:  Merck
CAS nummer:  
Testkits, chlortest, til Spectroquant®, Supelco®
Testkits, chlortest, til Spectroquant®, Supelco®
Klorreagens Cl₂-2*4, -, Prove, Nova 60, Multy, Move, Picco, 400 Tests
CAS nummer:  
Price
468,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request). Spectroquant® cell test kits.
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Spectroquant® reagent test kits
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*4 Reagenser til analyse af frit chlor er reagens Cl₂-1 1.00086.0001 og reagens Cl₂-2 1.00087.0001 sammen med tom kuvette og tilbehør 1.00089.0001
Reagenser til total chlor er reagens Cl₂-1 1.00086.0001, reagens Cl₂-2 1.00087.0001 og reagens Cl₂-3 1.00088.0001 sammen med tom kuvette og tilbehør 1.00089.0001
*7 Denne metode er officielt anerkendt af USEPA som en alternativ metode til undersøgelse af følgende:
*7-1 spildevand
*7-2 drikkevand
*7-3 hhv. drikke- og spildevand
Specifikationer
Specifikationer
  • Pakn:
    400 Tests
  • Beskrivelse:
    Klorreagens Cl₂-2*<sup>4</sup>
  • Til enhed:
    Prove, Nova 60, Multy, Move, Picco
  • Område:
    -
  • Cat. No.:
    1.00087.0001
  • M-Clarity quality level:
    MQ100
Specifikationer
Accessories
Produktinformation
  • Katalog #
    Til enhed
    Pakn
    Tilgængelighed
    Price
  • Chlorine, cells and accessories for photometric Cl₂ measurement*4
    Chlorine, cells and accessories for photometric Cl₂ measurement*4
    1.00089.0001
    Prove, Nova 60
    25
    Log ind for tilgængelighed
    1 * 25 Items - 748,00 kr