Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
o-Cresol ≥98%

o-Cresol ≥98%

Glass bottle for solids, 2.500 g
Katalog # A12307.0E
Giftig
Ætsende
Leverandør:   Thermo Fisher Scientific
o-Cresol ≥98%
Katalog # A12307.0E
CAS nummer:  95-48-7
o-Cresol ≥98%
o-Cresol ≥98%

Glass bottle for solids, 2.500 g

CAS nummer:  95-48-7
Price
Spørg for pris
Log ind for tilgængelighed
1 * 2.500 g
Product Details & Documents

Purity: 98+% CAS No.: 95-48-7 Molecular Formula: C7H8O Formula Weight: 108.14 Flash Point: 81°(177°F) Odour: Phenol-like Boiling Point: 190-192° Beilstein Reference: 506917 Merck Reference: 14,2579 UN No.: 3455 MDL No.: MFCD00002226

Synonymer:  o-Methylphenol
  • CAS nummer:95-48-7
  • Formel:C₇H₈O
  • MDL nummer:MFCD00002226
  • Molecular Weight:108.14 g/mol
  • Merck Index:14,2579
  • Kogende Pt:190…192 °C
  • Smeltende Pt:30…34 °C
  • Tæthed:1,048
  • Flash Pt:81 °C (177 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:202-423-8
  • FN:3455
  • ADR:6.1,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    2.500 g

Specifikationer