Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Tetrachlorethylen ≥99%

Glass bottle, 2.500 mL
Katalog # B20089.0F
Sundhed
Miljø
Leverandør:   Thermo Fisher Scientific
Tetrachlorethylen ≥99%
Katalog # B20089.0F
CAS nummer:  127-18-4
Tetrachlorethylen ≥99%
Tetrachlorethylen ≥99%

Glass bottle, 2.500 mL

CAS nummer:  127-18-4
Price
893,00 kr
Log ind for tilgængelighed
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 127-18-4 Molecular Formula: C2Cl4 Formula Weight: 165.83 Refractive Index: 1.5055 Odour: Ether-like Boiling Point: 120-121° Beilstein Reference: 1361721 Merck Reference: 14,9190 UN No.: 1897 MDL No.: MFCD00000834

Synonymer:  Tetrachloroethene, Ethylene tetrachloride, PCE, Perchloroethylene
  • CAS nummer:127-18-4
  • Formel:C₂Cl₄
  • MDL nummer:MFCD00000834
  • Molecular Weight:165.83 g/mol
  • Merck Index:14,9190
  • Kogende Pt:120…121 °C
  • Smeltende Pt:–22 °C
  • Tæthed:1,623
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:204-825-9
  • FN:1897
  • ADR:6.1,III
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2.500 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.