Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Acetonitril ≥99%

Glass bottle, 500 mL
Katalog # A19862.AP
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
Acetonitril ≥99%
Katalog # A19862.AP
CAS nummer:  75-05-8
Acetonitril ≥99%
Acetonitril ≥99%

Glass bottle, 500 mL

CAS nummer:  75-05-8
Price
659,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Purity: 99% CAS No.: 75-05-8 Molecular Formula: C2H3N Formula Weight: 41.05 Flash Point: 5°(41°F) Refractive Index: 1.3440 Odour: Pungent Melting Point: -48° Boiling Point: 80-82° Beilstein Reference: 741857 Merck Reference: 14,70 UN No.: 1648 MDL No.: MFCD00001878

Synonymer:  Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
  • CAS nummer:75-05-8
  • Formel:H₃CCN
  • MDL nummer:MFCD00001878
  • Molecular Weight:41.05 g/mol
  • Merck Index:14,70
  • Kogende Pt:80…82 °C
  • Smeltende Pt:–48 °C
  • Tæthed:0,786
  • Flash Pt:5 °C (41 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:200-835-2
  • FN:1648
  • ADR:3,II
  • NÅ:01-2119471307-38
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer