Acetonitril ≥99%
Glass bottle, 2.500 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 75-05-8 Molecular Formula: C2H3N Formula Weight: 41.05 Flash Point: 5°(41°F) Refractive Index: 1.3440 Odour: Pungent Melting Point: -48° Boiling Point: 80-82° Beilstein Reference: 741857 Merck Reference: 14,70 UN No.: 1648 MDL No.: MFCD00001878
Synonymer: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile
- CAS nummer:75-05-8
- Formel:H₃CCN
- MDL nummer:MFCD00001878
- Molecular Weight:41.05 g/mol
- Merck Index:14,70
- Kogende Pt:80…82 °C
- Smeltende Pt:–48 °C
- Tæthed:0,786
- Flash Pt:5 °C (41 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:200-835-2
- FN:1648
- ADR:3,II
- NÅ:01-2119471307-38
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:2.500 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



