Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Isooctan ≥99%

Glass bottle, 500 mL
Katalog # A19756.AP
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Thermo Fisher Scientific
Isooctan ≥99%
Katalog # A19756.AP
CAS nummer:  540-84-1
Isooctan ≥99%
Isooctan ≥99%

Glass bottle, 500 mL

CAS nummer:  540-84-1
Price
400,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Purity: 99% CAS No.: 540-84-1 Molecular Formula: C8H18 Formula Weight: 114.23 Flash Point: -7°(19°F) Refractive Index: 1.3910 Odour: Nearly Odourless Melting Point: -107° Boiling Point: 98-99° Beilstein Reference: 1696876 Merck Reference: 14,5193 UN No.: 1262 MDL No.: MFCD00008943

Synonymer:  Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
  • CAS nummer:540-84-1
  • Formel:(CH₃)₂CHCH₂C(CH₃)₃
  • MDL nummer:MFCD00008943
  • Molecular Weight:114.23 g/mol
  • Merck Index:14,5193
  • Kogende Pt:98…99 °C
  • Smeltende Pt:–107 °C
  • Tæthed:0,692
  • Flash Pt:–7 °C (19 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-759-1
  • FN:1262
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer