Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Isooctan ≥99%

Isooctan ≥99%

Glass bottle, 2.500 mL
Katalog # A19756.0F
Brandfarlig
Sundhed
Irriterende
Miljø
Leverandør:   Thermo Fisher Scientific
Isooctan ≥99%
Katalog # A19756.0F
CAS nummer:  540-84-1
Isooctan ≥99%
Isooctan ≥99%

Glass bottle, 2.500 mL

CAS nummer:  540-84-1
Price
931,00 kr
Log ind for tilgængelighed
1 * 2.500 mL
Product Details & Documents

Purity: 99% CAS No.: 540-84-1 Molecular Formula: C8H18 Formula Weight: 114.23 Flash Point: -7°(19°F) Refractive Index: 1.3910 Odour: Nearly Odourless Melting Point: -107° Boiling Point: 98-99° Beilstein Reference: 1696876 Merck Reference: 14,5193 UN No.: 1262 MDL No.: MFCD00008943

Synonymer:  Isobutyltrimethylmethane, Isooctane, Isobutyltrimethylethane
  • CAS nummer:540-84-1
  • Formel:(CH₃)₂CHCH₂C(CH₃)₃
  • MDL nummer:MFCD00008943
  • Molecular Weight:114.23 g/mol
  • Merck Index:14,5193
  • Kogende Pt:98…99 °C
  • Smeltende Pt:–107 °C
  • Tæthed:0,692
  • Flash Pt:–7 °C (19 °F)
  • Opbevaringstemperatur:Stue temperatur
  • EINECS:208-759-1
  • FN:1262
  • ADR:3,II
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    2.500 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.