Spermidin ≥99%
Glass bottle for solids, 5 g
Purity: 99% CAS No.: 124-20-9 Molecular Formula: H2N(CH2)3NH(CH2)4NH2 Formula Weight: 145.25 Flash Point: 110°(230°F) Refractive Index: 1.4790 Melting Point: 22-25° Boiling Point: 128-130°/14mm Beilstein Reference: 1698591 Merck Reference: 14,8742 UN No.: 2735 MDL No.: MFCD00008229
: N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
- :124-20-9
- :C₇H₁₉N₃
- :MFCD00008229
- :145.25 g/mol
- :14,8742
- :128…130 °C (14 mmHg)
- :22…25 °C
- :0,925
- :>110 °C (230 °F)
- :Stue temperatur
- :204-689-0
- :8,III
- Pakning:Glass bottle for solids
- Pakn:5 g