Orders LinkContactUsLinkComponent
Spermidin ≥99%
Glass bottle for solids, 5 g
  A19096.06
Spermidin ≥99%
  A19096.06
 :  124-20-9
Spermidin ≥99%
Spermidin ≥99%
Glass bottle for solids, 5 g
 :  124-20-9
1.510,00 kr
1 * 5 g
Purity: 99% CAS No.: 124-20-9 Molecular Formula: H2N(CH2)3NH(CH2)4NH2 Formula Weight: 145.25 Flash Point: 110°(230°F) Refractive Index: 1.4790 Melting Point: 22-25° Boiling Point: 128-130°/14mm Beilstein Reference: 1698591 Merck Reference: 14,8742 UN No.: 2735 MDL No.: MFCD00008229
 :  N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
  •  :124-20-9
  •  :C₇H₁₉N₃
  •  :MFCD00008229
  •  :145.25 g/mol
  •  :14,8742
  •  :128…130 °C (14 mmHg)
  •  :22…25 °C
  •  :0,925
  •  :>110 °C (230 °F)
  •  :Stue temperatur
  •  :204-689-0
  •  :8,III
  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g