(±)-2-Butanol ≥99%
Glass bottle, 2.500 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Purity: 99% CAS No.: 78-92-2 Molecular Formula: CH3CH2CH(OH)CH3 Formula Weight: 74.12 Flash Point: 26°(79°F) Refractive Index: 1.3970 Odour: Sweet Melting Point: -115° Boiling Point: 98-101° Beilstein Reference: 1718765 Merck Reference: 14,1541 UN No.: 1120 MDL No.: MFCD00004569
Synonymer: (±)-sec-Butanol, (±)-sec-Butyl alcohol, (±)-Ethyl methyl carbinol, (±)-2-Hydroxy butane, sec-Butanol, sec-Butyl alcohol, (±)-Methyl ethyl carbinol
- CAS nummer:78-92-2
- Formel:H₃CCH₂CH(OH)CH₃
- MDL nummer:MFCD00004569
- Molecular Weight:74.12 g/mol
- Merck Index:14,1541
- Kogende Pt:98…101 °C
- Smeltende Pt:–115 °C
- Tæthed:0,808
- Flash Pt:26 °C (79 °F)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-158-5
- FN:1120
- ADR:3,III
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:2.500 mL
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



