Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Styren ≥99% stabilised

Styren ≥99% stabilised

Glass bottle, 500 mL
Katalog # A18481.AP
Sundhed
Brandfarlig
Irriterende
Leverandør:   Thermo Fisher Scientific
Styren ≥99% stabilised
Katalog # A18481.AP
CAS nummer:  100-42-5
Styren ≥99% stabilised
Styren ≥99% stabilised

Glass bottle, 500 mL

CAS nummer:  100-42-5
Price
376,00 kr
Log ind for tilgængelighed
1 * 500 mL
Product Details & Documents

Stabilized with 10-15 ppm 4-tert-butylcatechol

Purity: 99% CAS No.: 100-42-5 Molecular Formula: C8H8 Formula Weight: 104.15 Storage Temperature: 2°C to 8°C Flash Point: 31°(88°F) Refractive Index: 1.5460 Odour: Pungent pH: neutral Melting Point: -31° Boiling Point: 145-146° Beilstein Reference: 1071236 Merck Reference: 14,8860 UN No.: 2055 MDL No.: MFCD00008612

Synonymer:  Phenylethene, Phenylethylene, Vinylbenzene
  • CAS nummer:100-42-5
  • Formel:C₈H₈
  • MDL nummer:MFCD00008612
  • Molecular Weight:104.15 g/mol
  • Merck Index:14,8860
  • Kogende Pt:145…146 °C
  • Smeltende Pt:–31 °C
  • Tæthed:0,906
  • Flash Pt:31 °C (88 °F)
  • Opbevaringstemperatur:Køleskab
  • FN:2055
  • ADR:3,III
  •  :stabiliseret med 4-tert-Butylpyrocatechol 10-15 ppm
  • EINECS:202-851-5
Specifikationer

Specifikationer

  • Pakning:
    Glass bottle
  • Pakn:
    500 mL

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.