Styren ≥99% stabilised
Glass bottle, 100 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Stabilized with 10-15 ppm 4-tert-butylcatechol
Purity: 99% CAS No.: 100-42-5 Molecular Formula: C8H8 Formula Weight: 104.15 Storage Temperature: 2°C to 8°C Flash Point: 31°(88°F) Refractive Index: 1.5460 Odour: Pungent pH: neutral Melting Point: -31° Boiling Point: 145-146° Beilstein Reference: 1071236 Merck Reference: 14,8860 UN No.: 2055 MDL No.: MFCD00008612
Synonymer: Phenylethene, Phenylethylene, Vinylbenzene
- CAS nummer:100-42-5
- Formel:C₈H₈
- MDL nummer:MFCD00008612
- Molecular Weight:104.15 g/mol
- Merck Index:14,8860
- Kogende Pt:145…146 °C
- Smeltende Pt:–31 °C
- Tæthed:0,906
- Flash Pt:31 °C (88 °F)
- Opbevaringstemperatur:Køleskab
- FN:2055
- ADR:3,III
- :stabiliseret med 4-tert-Butylpyrocatechol 10-15 ppm
- EINECS:202-851-5
Documents
Specifikationer
Specifikationer
- Pakning:Glass bottle
- Pakn:100 mL



